Learn More
Medchemexpress LLC Annexin A1/ANXA1 Mo 1mg
SDP
Catalog No. 5000195464
Encompass_Preferred
Annexin A1 (ANXA1) Protein, Mouse (His) is a full-length recombinant mouse annexin A1 expressed in Escherichia coli with an N-terminal 6xHis tag, supplied as a lyophilized powder. The protein is >95% pure, has an approximate molecular weight of 40 kDa, and low endotoxin (<1 EU/μg); it is formulated with trehalose for stability and intended for in vitro studies of inflammation, immune signaling, phagocytosis, and chemotaxis.
  • Full-length recombinant protein with N-terminal His tag.
  • Expressed in Escherichia coli for scalable production.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximately 40 kDa molecular weight.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Demonstrated biological activity in suppressing TNF-alpha secretion in RAW264.7 cells.
  • Multiple size options available to fit experimental needs.
Catalog No. 50-001-95464 Supplier Medchemexpress LLC Supplier No. HYP720781MG
Annexin A1 (ANXA1) Protein, Mouse (His) is a full-length recombinant mouse annexin A1 expressed in Escherichia coli with an N-terminal 6xHis tag, supplied as a lyophilized powder. The protein is >95% pure, has an approximate molecular weight of 40 kDa, and low endotoxin (<1 EU/μg); it is formulated with trehalose for stability and intended for in vitro studies of inflammation, immune signaling, phagocytosis, and chemotaxis.
  • Full-length recombinant protein with N-terminal His tag.
  • Expressed in Escherichia coli for scalable production.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximately 40 kDa molecular weight.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Demonstrated biological activity in suppressing TNF-alpha secretion in RAW264.7 cells.
  • Multiple size options available to fit experimental needs.