Learn More
Medchemexpress LLC Annexin A1/ANXA1 Mo 20ug
SDP
Catalog No. 5000195923
Encompass_Preferred
Recombinant mouse Annexin A1 (ANXA1) is supplied lyophilized as an N-terminal His-tagged protein expressed in Pichia pastoris for research applications in calcium-dependent phospholipid binding, inflammation, and Annexin A1 functional studies.
  • Expressed in Pichia pastoris with N-terminal His tag.
  • Approximately 42 kDa molecular weight.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized from PBS with 6% trehalose, pH 7.4.
  • Recommended reconstitution concentration ≥100 μg/mL.
  • Stable when stored at -20°C; aliquot and store at -80°C for long-term.
  • Available in research sizes (20 μg, 50 μg, 100 μg) and larger quantities on request.
Catalog No. 50-001-95923 Supplier Medchemexpress LLC Supplier No. HYP7171320UG
Recombinant mouse Annexin A1 (ANXA1) is supplied lyophilized as an N-terminal His-tagged protein expressed in Pichia pastoris for research applications in calcium-dependent phospholipid binding, inflammation, and Annexin A1 functional studies.
  • Expressed in Pichia pastoris with N-terminal His tag.
  • Approximately 42 kDa molecular weight.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized from PBS with 6% trehalose, pH 7.4.
  • Recommended reconstitution concentration ≥100 μg/mL.
  • Stable when stored at -20°C; aliquot and store at -80°C for long-term.
  • Available in research sizes (20 μg, 50 μg, 100 μg) and larger quantities on request.