Learn More
Medchemexpress LLC Annexin A1/ANXA1 Mo 50ug
SDP
Catalog No. 5000195526
Encompass_Preferred
Recombinant mouse Annexin A1 expressed in Pichia pastoris with an N-terminal His tag, supplied as a lyophilized protein. ANXA1 participates in innate and adaptive immune responses, inflammation resolution, and Ca2+-dependent phospholipid membrane interactions, making this reagent useful for biochemical characterization and cell biology studies. The product is supplied at 50 μg, exhibits >90% purity by reducing SDS-PAGE, and has an approximate molecular weight of 42 kDa.
Catalog No. 50-001-95526 Supplier Medchemexpress LLC Supplier No. HYP7171350UG
Recombinant mouse Annexin A1 expressed in Pichia pastoris with an N-terminal His tag, supplied as a lyophilized protein. ANXA1 participates in innate and adaptive immune responses, inflammation resolution, and Ca2+-dependent phospholipid membrane interactions, making this reagent useful for biochemical characterization and cell biology studies. The product is supplied at 50 μg, exhibits >90% purity by reducing SDS-PAGE, and has an approximate molecular weight of 42 kDa.