Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A1/ANXA1 Mo 5ug
SDP

Catalog No. 5000196276
Encompass_Preferred

Recombinant mouse Annexin A1 (ANXA1) protein expressed in E. coli with an N-terminal 6xHis tag. Purified to >95% by reducing SDS-PAGE and supplied in small pack sizes for research use; suitable for biochemical assays, antibody validation, and functional studies. Store frozen and reconstitute per manufacturer recommendations.

  • Recombinant mouse ANXA1 protein with N-terminal 6xHis tag.
  • Expressed in E. coli for efficient production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 40 kDa.
  • Suitable for biochemical assays and antibody validation.
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for extended storage.

Catalog No. 50-001-96276 Supplier Medchemexpress LLC Supplier No. HYP720785UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse Annexin A1 (ANXA1) protein expressed in E. coli with an N-terminal 6xHis tag. Purified to >95% by reducing SDS-PAGE and supplied in small pack sizes for research use; suitable for biochemical assays, antibody validation, and functional studies. Store frozen and reconstitute per manufacturer recommendations.

  • Recombinant mouse ANXA1 protein with N-terminal 6xHis tag.
  • Expressed in E. coli for efficient production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 40 kDa.
  • Suitable for biochemical assays and antibody validation.
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.