Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A1 (ANXA1) protein, human | >95.0% | 5 UG
SDP

Catalog No. 5000291542
Encompass_Preferred

Recombinant human Annexin A1 protein expressed in E. coli and supplied lyophilized. This tag free protein is intended for biochemical and immunological assays; a certificate of analysis and handling/storage guidance are provided to support experimental reproducibility.

  • Recombinant human Annexin A1, tag free
  • Purity greater than 95% by reducing SDS-PAGE
  • Molecular weight approximately 34-39 kDa
  • Lyophilized formulation from buffered salts with optional 8% trehalose
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C for extended storage

Catalog No. 50-002-91542 Supplier Medchemexpress LLC Supplier No. HYP75125UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Annexin A1 protein expressed in E. coli and supplied lyophilized. This tag free protein is intended for biochemical and immunological assays; a certificate of analysis and handling/storage guidance are provided to support experimental reproducibility.

  • Recombinant human Annexin A1, tag free
  • Purity greater than 95% by reducing SDS-PAGE
  • Molecular weight approximately 34-39 kDa
  • Lyophilized formulation from buffered salts with optional 8% trehalose
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C for extended storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.