Learn More
Medchemexpress LLC Annexin A1/ANXA1 protein, human | ≥ 95% | 34-39 kDa | 100 UG
SDP
Catalog No. 5000399272
Encompass_Preferred
Annexin A1 protein, human, is a Ca2+-dependent phospholipid binding protein located on the cytosolic face of the plasma membrane. It plays multifunctional roles in innate and adaptive immunity, primarily regulating inflammatory cells and inflammation resolution by inhibiting neutrophil transmigration and recruitment at inflammatory sites. This protein also inhibits phospholipase A2 activity and binds 2-4 calcium ions with high affinity.
  • Inhibits phospholipase A2 activity
  • Binds 2-4 calcium ions with high affinity
  • Regulates inflammatory cells and inflammation resolution
  • Inhibits neutrophil transmigration and recruitment
Catalog No. 50-003-99272 Supplier Medchemexpress LLC Supplier No. HYP7512100UG
Annexin A1 protein, human, is a Ca2+-dependent phospholipid binding protein located on the cytosolic face of the plasma membrane. It plays multifunctional roles in innate and adaptive immunity, primarily regulating inflammatory cells and inflammation resolution by inhibiting neutrophil transmigration and recruitment at inflammatory sites. This protein also inhibits phospholipase A2 activity and binds 2-4 calcium ions with high affinity.
  • Inhibits phospholipase A2 activity
  • Binds 2-4 calcium ions with high affinity
  • Regulates inflammatory cells and inflammation resolution
  • Inhibits neutrophil transmigration and recruitment