Learn More
Medchemexpress LLC Annexin A2/ANXA2 Mo 20ug | 20ug
SDP
Catalog No. 5000187203
Encompass_Preferred
Annexin A2/ANXA2 protein is a calcium-regulated membrane-binding protein that binds two calcium ions Annexin A2/ANXA2 Protein Mouse (His) is the recombinant mouse-derived Annexin A2/ANXA2 protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-87203 Supplier Medchemexpress LLC Supplier No. HYP7756120UG
Annexin A2/ANXA2 protein is a calcium-regulated membrane-binding protein that binds two calcium ions Annexin A2/ANXA2 Protein Mouse (His) is the recombinant mouse-derived Annexin A2/ANXA2 protein expressed by E coli with N-His labeled tag