Learn More
Bioss Annexin A2 - C-terminal region
SDP
Catalog No. 5000065152
Encompass_Preferred
Annexin A2 (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Chicken Clonality Monoclonal Applications WB IP IHC M298Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65152 Supplier Bioss Supplier No. BSM70336M
Annexin A2 (C-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Chicken Clonality Monoclonal Applications WB IP IHC M298Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol