Learn More
Medchemexpress LLC Annexin A4/ANXA4 Mo 50ug
SDP
Catalog No. 5000196308
Encompass_Preferred
Recombinant mouse Annexin A4 (ANXA4) is supplied as a lyophilized powder with N-terminal 10xHis and C-terminal Myc tags, produced in E. coli. The protein is designed for biochemical assays, antibody validation, structural studies, and investigations of membrane dynamics and exocytosis. It is provided with high purity and low endotoxin, formulated in a Tris-HCl/NaCl/trehalose buffer, and should be aliquoted and frozen for long-term storage.
  • Lyophilized recombinant protein for flexible reconstitution.
  • N-terminal His and C-terminal Myc tags for detection and purification.
  • Greater than 90% purity by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Formulated in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Recommended storage at -20°C with aliquots at -80°C for extended stability.
  • Available in multiple sizes, including 50 μg.
Catalog No. 50-001-96308 Supplier Medchemexpress LLC Supplier No. HYP7207950UG
Add to Cart
Add to Cart
Recombinant mouse Annexin A4 (ANXA4) is supplied as a lyophilized powder with N-terminal 10xHis and C-terminal Myc tags, produced in E. coli. The protein is designed for biochemical assays, antibody validation, structural studies, and investigations of membrane dynamics and exocytosis. It is provided with high purity and low endotoxin, formulated in a Tris-HCl/NaCl/trehalose buffer, and should be aliquoted and frozen for long-term storage.
  • Lyophilized recombinant protein for flexible reconstitution.
  • N-terminal His and C-terminal Myc tags for detection and purification.
  • Greater than 90% purity by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Formulated in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Recommended storage at -20°C with aliquots at -80°C for extended stability.
  • Available in multiple sizes, including 50 μg.