Learn More
Medchemexpress LLC Annexin A4/ANXA4 Mo 5ug
SDP
Catalog No. 5000195575
Encompass_Preferred
Recombinant mouse Annexin A4 (ANXA4) fragment (residues E2-D319) supplied as a lyophilized His-Myc-tagged protein for biochemical and molecular biology research. Expressed in E. coli and purified to greater than 90% purity, the product is formulated for stability in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0, and is intended for research use only.
  • Recombinant fragment spanning residues E2-D319.
  • N-terminal 10xHis and C-terminal Myc tags for detection and purification.
  • Expressed in E. coli for high-yield production.
  • Purified to greater than 90% purity by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Stable when stored at -20°C; reconstituted material stable at 4°C for one week.
  • Available in small research sizes, including 5 μg.
Catalog No. 50-001-95575 Supplier Medchemexpress LLC Supplier No. HYP720795UG
Recombinant mouse Annexin A4 (ANXA4) fragment (residues E2-D319) supplied as a lyophilized His-Myc-tagged protein for biochemical and molecular biology research. Expressed in E. coli and purified to greater than 90% purity, the product is formulated for stability in 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0, and is intended for research use only.
  • Recombinant fragment spanning residues E2-D319.
  • N-terminal 10xHis and C-terminal Myc tags for detection and purification.
  • Expressed in E. coli for high-yield production.
  • Purified to greater than 90% purity by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Stable when stored at -20°C; reconstituted material stable at 4°C for one week.
  • Available in small research sizes, including 5 μg.