Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A5/ANXA5 protein, mouse (His, Myc) | >95.0% | 100UG
SDP

Catalog No. 5000176913
Encompass_Preferred

Recombinant mouse Annexin A5 (ANXA5) is a His- and Myc-tagged protein supplied as a lyophilized powder for biochemical and cell biology applications. It binds negatively charged phospholipids in a calcium-dependent manner and is commonly used to detect apoptosis, study membrane binding, and promote vesicle aggregation.

  • His- and Myc-tagged recombinant protein for easy detection and purification.
  • Produced in Escherichia coli and supplied as lyophilized powder for stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Formulated with Tris-HCl, NaCl, and trehalose to preserve activity on drying.
  • Useful for apoptosis assays, phospholipid binding studies, and membrane fusion experiments.
  • Store at -20°C; after reconstitution store at 4°C short term or freeze aliquots for long-term storage.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O to maintain integrity.

Catalog No. 50-001-76913 Supplier Medchemexpress LLC Supplier No. HYP702573100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Annexin A5 (ANXA5) is a His- and Myc-tagged protein supplied as a lyophilized powder for biochemical and cell biology applications. It binds negatively charged phospholipids in a calcium-dependent manner and is commonly used to detect apoptosis, study membrane binding, and promote vesicle aggregation.

  • His- and Myc-tagged recombinant protein for easy detection and purification.
  • Produced in Escherichia coli and supplied as lyophilized powder for stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Formulated with Tris-HCl, NaCl, and trehalose to preserve activity on drying.
  • Useful for apoptosis assays, phospholipid binding studies, and membrane fusion experiments.
  • Store at -20°C; after reconstitution store at 4°C short term or freeze aliquots for long-term storage.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O to maintain integrity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.