Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A5/ANXA5 protein, human (solution) | >95.0% | 10 UG
SDP

Catalog No. 5000292894
Encompass_Preferred

Recombinant human Annexin A5 (ANXA5) is a calcium-dependent phospholipid-binding protein supplied lyophilized for research use. It is suitable for apoptosis detection, coagulation studies, and in vitro biochemical assays, and is provided with activity and handling recommendations to support experimental workflows.

  • Recombinant human protein, approx. 33.0 kDa.
  • Purity >95.0% by reducing SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, containing 8% trehalose.
  • Do not reconstitute below 100 μg/mL; aliquot and freeze for long-term storage.
  • Activity: ED50 = 0.1537 μg/mL (alkaline phosphatase induction assay).
  • Intended for in vitro research applications such as apoptosis and coagulation assays.

Catalog No. 50-002-92894 Supplier Medchemexpress LLC Supplier No. HYP7517A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Annexin A5 (ANXA5) is a calcium-dependent phospholipid-binding protein supplied lyophilized for research use. It is suitable for apoptosis detection, coagulation studies, and in vitro biochemical assays, and is provided with activity and handling recommendations to support experimental workflows.

  • Recombinant human protein, approx. 33.0 kDa.
  • Purity >95.0% by reducing SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, containing 8% trehalose.
  • Do not reconstitute below 100 μg/mL; aliquot and freeze for long-term storage.
  • Activity: ED50 = 0.1537 μg/mL (alkaline phosphatase induction assay).
  • Intended for in vitro research applications such as apoptosis and coagulation assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.