Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A5 protein, human | >95.0% | 50 UG
SDP

Catalog No. 5000293488
Encompass_Preferred

Recombinant human Annexin A5 is a full-length, tag-free protein supplied as a lyophilized powder for biochemical and cell biology applications such as apoptosis detection and coagulation studies. The protein is expressed in E. coli and characterized for purity, activity, and low endotoxin, providing a reliable reagent for in vitro assays.

  • Full-length, tag free recombinant human protein.
  • Lyophilized powder formulation in PBS with 8% trehalose.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight approximately 33.0 kDa.
  • Endotoxin <1 EU/μg by LAL method.
  • Stable frozen; store at -20°C and freeze aliquots for long-term storage.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Demonstrated biological activity (ED50 0.1537 μg/mL in an alkaline phosphatase assay).

Catalog No. 50-002-93488 Supplier Medchemexpress LLC Supplier No. HYP7517A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human Annexin A5 is a full-length, tag-free protein supplied as a lyophilized powder for biochemical and cell biology applications such as apoptosis detection and coagulation studies. The protein is expressed in E. coli and characterized for purity, activity, and low endotoxin, providing a reliable reagent for in vitro assays.

  • Full-length, tag free recombinant human protein.
  • Lyophilized powder formulation in PBS with 8% trehalose.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight approximately 33.0 kDa.
  • Endotoxin <1 EU/μg by LAL method.
  • Stable frozen; store at -20°C and freeze aliquots for long-term storage.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Demonstrated biological activity (ED50 0.1537 μg/mL in an alkaline phosphatase assay).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.