Learn More
ABclonal Technology Annexin A6/Annexin VI
SDP
Supplier:  ABclonal Technology A22038100UL
Encompass_Preferred
Annexin VI belongs to a family of calcium-dependent membrane and phospholipid binding proteins Several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways The annexin VI gene is approximately 60 kbp long and contains 26 exons It encodes a protein of about 68 kDa that consists of eight 68-amino acid repeats separated by linking sequences of variable lengths It is highly similar to human annexins I and II sequences each of which contain four such repeats Annexin VI has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis Alternatively spliced transcript variants have been described
Catalog No. 50-000-07516