Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A8/ANXA8 protein, human (His) | >95.0% | 100 UG
SDP

Catalog No. 5000293162
Encompass_Preferred

Recombinant human Annexin A8 is a full-length, His-tagged protein expressed in E. coli and supplied lyophilized for research use. Annexin A8 acts as an anticoagulant by indirectly inhibiting the thromboplastin-specific complex. The product is provided with high purity and low endotoxin, suitable for biochemical and functional studies.

  • Species: human
  • Expression system: E. coli
  • Protein length: full-length isoform 1
  • Molecular weight: ≈ 38 kDa
  • Tag: N-6 His
  • Purity: >95.0%
  • Endotoxin: <1 EU/μg
  • Formulation: lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4
  • Storage: store lyophilized at -20°C; freeze aliquots after reconstitution

Catalog No. 50-002-93162 Supplier Medchemexpress LLC Supplier No. HYP75505100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human Annexin A8 is a full-length, His-tagged protein expressed in E. coli and supplied lyophilized for research use. Annexin A8 acts as an anticoagulant by indirectly inhibiting the thromboplastin-specific complex. The product is provided with high purity and low endotoxin, suitable for biochemical and functional studies.

  • Species: human
  • Expression system: E. coli
  • Protein length: full-length isoform 1
  • Molecular weight: ≈ 38 kDa
  • Tag: N-6 His
  • Purity: >95.0%
  • Endotoxin: <1 EU/μg
  • Formulation: lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4
  • Storage: store lyophilized at -20°C; freeze aliquots after reconstitution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.