Learn More
STEMCELL Technologies Annexin V Binding Buffer, Size: 50 mL
SDP
Catalog No. 502390344
Encompass
Annexin V Binding Buffer is an isotonic buffer used in conjunction with Annexin V dyes (e.g. Catalog 100-0329) and either 7-AAD (Catalog 75001) or Propidium Iodide (Catalog 75002). This buffer contains an optimal concentration of calcium that facilitates the binding of Annexin V to phosphatidylserine on the cell surface which is essential for the detection of early-stage cell apoptosis by flow cytometry.
Catalog No. 50-239-0344 Supplier STEMCELL Technologies Supplier No. 1000334
Annexin V Binding Buffer is an isotonic buffer used in conjunction with Annexin V dyes (e.g. Catalog 100-0329) and either 7-AAD (Catalog 75001) or Propidium Iodide (Catalog 75002). This buffer contains an optimal concentration of calcium that facilitates the binding of Annexin V to phosphatidylserine on the cell surface which is essential for the detection of early-stage cell apoptosis by flow cytometry.