Learn More
Biotium Annexin V, Biotin Conjugate
Catalog No. 501964418
Encompass_Preferred
Annexin V fluorescent conjugates can be used to detect apoptotic cells. The human anticoagulant Annexin V is a 35-36 kDa calcium-dependent phospholipid-binding protein with high affinity for phosphatidylserine (PS). In normal viable cells, PS is located on the inner leaflet of the cytoplasmic membrane. In apoptotic cells, however, PS is translocated from the inner to the outer leaflet of the plasma membrane, where it can be detected by fluorescently labeled Annexin V. Biotin Annexin V conjugate is provided at 50 ug/mL in buffer with azide as a preservative.
Catalog No. 50-196-4418 Supplier Biotium Supplier No. 29013
Annexin V fluorescent conjugates can be used to detect apoptotic cells. The human anticoagulant Annexin V is a 35-36 kDa calcium-dependent phospholipid-binding protein with high affinity for phosphatidylserine (PS). In normal viable cells, PS is located on the inner leaflet of the cytoplasmic membrane. In apoptotic cells, however, PS is translocated from the inner to the outer leaflet of the plasma membrane, where it can be detected by fluorescently labeled Annexin V. Biotin Annexin V conjugate is provided at 50 ug/mL in buffer with azide as a preservative.