Learn More
Apexbio Technology LLC Annexin V-Biotin Apoptosis Kit, 25 assays
SDP
Supplier:  Apexbio Technology LLC K200925
Encompass_Preferred
Annexin V is a cellular protein, which plays important roles in the inhibition of the activity of phospholipase A1 and blood coagulation by competing for phosphatidylserine (PS) binding sites with prothrombin. Annexin V has a high affinity to PS and is used as a probe to detect cells that have expressed PS on the cell surface. After initiating apoptosis, cells translocate phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface soon, which can be easily detected by Annexin V.Annexin V-Biotin Apoptosis Detection Kit uses Biotin-conjugated Annexin V that can easily detect PS on the cell surface after initiating apoptosis. Annexin V-Biotin can be easily detected in combination with conventional dye-staining using any avidin- or streptavidine-dye reagents, such as (strept)avidine-peroxidase, -fluorescein, -alkaline phosphatase (AP), and --gal, etc. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8357