Learn More
Apexbio Technology LLC Annexin V-Cy3 Apoptosis Kit, 100 assays. Cas: MFCD: NA. Detects Apoptosis within 10 min.
SDP
Catalog No. 501998353
Encompass_Preferred
Annexin V is a cellular protein, which plays important roles in the inhibition of the activity of phospholipase A1 and blood coagulation by competing for phosphatidylserine (PS) binding sites with prothrombin. Annexin V has a high affinity to PS and is used as a probe to detect cells that have expressed PS on the cell surface. After initiating apoptosis, cells translocate phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface soon, which can be easily detected by Annexin V.Annexin V-Cy3 Apoptosis Detection Kit uses a fluorescent conjugate of Annexin V that can easily detect PS on the cell surface after initiating apoptosis. The one-step staining procedure needs only 10 minutes. The result can be analyzed by fluorescence microscopy or by flow cytometry. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8353 Supplier Apexbio Technology LLC Supplier No. K2004100
Annexin V is a cellular protein, which plays important roles in the inhibition of the activity of phospholipase A1 and blood coagulation by competing for phosphatidylserine (PS) binding sites with prothrombin. Annexin V has a high affinity to PS and is used as a probe to detect cells that have expressed PS on the cell surface. After initiating apoptosis, cells translocate phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface soon, which can be easily detected by Annexin V.Annexin V-Cy3 Apoptosis Detection Kit uses a fluorescent conjugate of Annexin V that can easily detect PS on the cell surface after initiating apoptosis. The one-step staining procedure needs only 10 minutes. The result can be analyzed by fluorescence microscopy or by flow cytometry. Other sizes are also available. Please inqury us for quote.