Learn More
Cell Signaling Technology Annexin V (E3W8V) Rabbit Monoclonal Antibody, Recombinant, Unconjugated, Human, Mouse, Rat, 100 μL
SDP
Catalog No. 502733144
Encompass
The Annexin V (E3W8V) Rabbit Monoclonal Antibody is a recombinant antibody engineered to recognize endogenous levels of total annexin V protein. Annexin V is an approximately 30 kDa protein that binds to phospholipids in a calcium-dependent manner and functions as an inhibitor of the protein kinase C pathway. Due to its robust and specific binding to phosphatidylserine, this antibody is a useful reagent for the detection of apoptotic cells.
  • Validated for use in western blotting, immunohistochemistry (paraffin), immunofluorescence (immunocytochemistry), and flow cytometry (fixed/permeabilized) applications.
  • Species reactivity includes human, mouse, and rat.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended for research use only; not for use in diagnostic procedures.
Catalog No. 50-273-3144 Supplier Cell Signaling Technology Supplier No. 54043S
The Annexin V (E3W8V) Rabbit Monoclonal Antibody is a recombinant antibody engineered to recognize endogenous levels of total annexin V protein. Annexin V is an approximately 30 kDa protein that binds to phospholipids in a calcium-dependent manner and functions as an inhibitor of the protein kinase C pathway. Due to its robust and specific binding to phosphatidylserine, this antibody is a useful reagent for the detection of apoptotic cells.
  • Validated for use in western blotting, immunohistochemistry (paraffin), immunofluorescence (immunocytochemistry), and flow cytometry (fixed/permeabilized) applications.
  • Species reactivity includes human, mouse, and rat.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended for research use only; not for use in diagnostic procedures.