Learn More
Enzo Life Sciences Annexin V-EGFP apoptosis detection kit (100 tests)
SDP
Catalog No. 502010139
Encompass_Preferred
EGFP (enhanced green fluorescent protein) has emerged as an ideal fluorochrome for FACS analysis and fluorescent microscopy as it offers a more intense signal than wild-type GFP. It has Ex(max): 488nm and Em(max): 530nm. The green signal produced makes EGFP ideal for dual staining with propidium iodide (PI). Applications: Flow Cytometry, Fluorescence microscopy .
Catalog No. 50-201-0139 Supplier Enzo Life Sciences Supplier No. ALX850253KI02
EGFP (enhanced green fluorescent protein) has emerged as an ideal fluorochrome for FACS analysis and fluorescent microscopy as it offers a more intense signal than wild-type GFP. It has Ex(max): 488nm and Em(max): 530nm. The green signal produced makes EGFP ideal for dual staining with propidium iodide (PI). Applications: Flow Cytometry, Fluorescence microscopy .