Learn More
Apexbio Technology LLC Annexin-V-EGFP-Apoptosis-Kit
SDP
Catalog No. 5000567604
Encompass_Preferred
Annexin V-EGFP Apoptosis Kit is commonly used in biomedical research to study apoptosis primarily in cell biology and oncology This kit originates from fluorescence-based methodologies and leverages the affinity of Annexin V for phosphatidylserine which is externalized on the surface of apoptotic cells It allows for the visualization of apoptotic processes in real-time under a microscope Commonly this assay is applied in vitro with activity observable at various concentration levels Typical applications include usage in numerous cell models to evaluate apoptosis assess drug efficacy and understand signaling pathways involved in cell death This tool is valuable for drug screening and mechanistic studies where experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67604 Supplier Apexbio Technology LLC Supplier No. K2006100
Annexin V-EGFP Apoptosis Kit is commonly used in biomedical research to study apoptosis primarily in cell biology and oncology This kit originates from fluorescence-based methodologies and leverages the affinity of Annexin V for phosphatidylserine which is externalized on the surface of apoptotic cells It allows for the visualization of apoptotic processes in real-time under a microscope Commonly this assay is applied in vitro with activity observable at various concentration levels Typical applications include usage in numerous cell models to evaluate apoptosis assess drug efficacy and understand signaling pathways involved in cell death This tool is valuable for drug screening and mechanistic studies where experimental concentrations typically depend on the specific research design