Learn More
Apexbio Technology LLC Annexin V-FITC Apoptosis Kit, 100Assays Cas No: NA MFCD: NA
SDP
Catalog No. NC1628148
Encompass_Preferred
Annexin V is a cellular protein, which plays important roles in the inhibition of the activity of phospholipase A1 and blood coagulation by competing for phosphatidylserine (PS) binding sites with prothrombin. Annexin V has a high affinity to PS and is used as a probe to detect cells that have expressed PS on the cell surface. After initiating apoptosis, cells translocate phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface soon, which can be easily detected by Annexin V. There are other sizes available. Please inqury us for quote.
Catalog No. NC1628148 Supplier Apexbio Technology LLC Supplier No. K2003100
Annexin V is a cellular protein, which plays important roles in the inhibition of the activity of phospholipase A1 and blood coagulation by competing for phosphatidylserine (PS) binding sites with prothrombin. Annexin V has a high affinity to PS and is used as a probe to detect cells that have expressed PS on the cell surface. After initiating apoptosis, cells translocate phosphatidylserine (PS) from the inner face of the plasma membrane to the cell surface soon, which can be easily detected by Annexin V. There are other sizes available. Please inqury us for quote.