Learn More
Enzo Life Sciences Annexin V-FITC apoptosis detection kit (300tests)
SDP
Catalog No. 502010093
Encompass_Preferred
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488nm, Em(max): 530nm). Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-0093 Supplier Enzo Life Sciences Supplier No. ALX850020KI01
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488nm, Em(max): 530nm). Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .