Learn More
Sigma Aldrich Fine Chemicals Biosciences Annexin V fITC Apoptosis D
Catalog No. 502905328
Encompass
Annexin V-fITC kit allows fluorescent detection of annexin V bound to apoptotic cells and quantitative determination by flow cytometry. The AnnexinV-fITC kit uses annexin V conjugated with fluorescein isothiocyante (fITC) to label phosphatidylserine sites on the membrane surface. The kit includes propidium iodide (PI) to label the cellular DNA in necrotic cells where the cell membrane has been totally compromised. This combination allows the differentiation among early apoptotic cells (annexin V positive PI negative) necrotic cells (annexin V positive PI positive) and viable cells (annexin V negative PI negative).
Catalog No. 50-290-5328 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. APOAF60TST
Annexin V-fITC kit allows fluorescent detection of annexin V bound to apoptotic cells and quantitative determination by flow cytometry. The AnnexinV-fITC kit uses annexin V conjugated with fluorescein isothiocyante (fITC) to label phosphatidylserine sites on the membrane surface. The kit includes propidium iodide (PI) to label the cellular DNA in necrotic cells where the cell membrane has been totally compromised. This combination allows the differentiation among early apoptotic cells (annexin V positive PI negative) necrotic cells (annexin V positive PI positive) and viable cells (annexin V negative PI negative).