Learn More
Invitrogen™ Annexin V Recombinant Protein, Biotin
Description
Description: Annexins are a family of calcium-dependent phospholipid-binding proteins, which bind to phosphatidylserine (PS) to identify apoptotic cells. In healthy cells, PS is predominantly located along the cytosolic side of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution in the phospholipid bilayer and translocates to the extracellular membrane, which is detectable with fluorescently labeled Annexin V. In early stages of apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI) and 7-AAD, therefore cells which display only Annexin V staining (PI/7-AAD negative) are in early stages of apoptosis. During late-stage apoptosis, loss of cell membrane integrity allows Annexin V binding to cytosolic PS, as well as cell uptake of PI and 7-AAD. Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry. Annexin V has also been identified as an anticoagulant protein in the blood coagulation cascade, by acting as an inhibitor of prothrombin activation. The presence of antibodies to Annexin V is associated with systemic lupus erythematosus (SLE), recurrent spontaneous abortions and systemic sclerosis (SSc). Purity: Annexin V is >89% pure as demonstrated by SDS-gel electrophoresis and reverse phase HPLC. Biotin labelling allows simple direct detection of PS by FACS analysis using fluorescence-labeled Streptavidin.
Specifications
Specifications
| Accession Number | P08758, P14668, P48036 |
| Concentration | 5 μL/Test |
| For Use With (Application) | Neutralization, Flow Cytometry |
| Formulation | TBS with 1% BSA and 0.02% sodium azide; pH 7.4 |
| Gene ID (Entrez) | 11747, 25673, 308 |
| Molecular Weight (g/mol) | 35.8kDa |
| Name | Annexin V |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 20 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.