Learn More
Enzo Life Sciences Annexin V (human), (recombinant) (FITC conjugate) (100tests)
SDP
Supplier:  Enzo Life Sciences ALX2092563100
Encompass_Preferred
FITC-labelled recombinant human annexin V fusion protein (produced in E. coli) shows bright green fluorescence (Ex(max): 488nm; Em(max): 530nm). FITC-labelled recombinant human annexin V fusion protein (produced in E. coli) shows bright green fluorescence (Ex(max): 488nm; Em(max): 530nm). Concentration: 1mg/ml. Purity: ≥90%. MW: ~52kDa (SDS-PAGE). Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide. Source: Produced in E.coli. Specificity: Binds to phosphatidylserine (PS). Long Term Storage: +4°C. Use/Stability: Stable for at least one year after receipt when stored at +4°C. Handling: Protect from light. Do not freeze. Applications: Flow Cytometry .
Catalog No. 50-200-8064