Learn More
Enzo Life Sciences Annexin V (human), (recombinant) (FITC conjugate) (300tests)
SDP
Supplier:  Enzo Life Sciences ALX209250T300
Encompass_Preferred
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488nm; Em(max): 530nm). FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488nm; Em(max): 530nm). Purity: ≥98% (SDS-PAGE, HPLC). MW: ~35.8kDa. Formulation: Liquid. In 50mM TRIS, pH 7.4, containing 100mM sodium chloride, 1% BSA and 0.02% sodium azide. Source: Produced in E. coli. Specificity: Binds to phosphatidylserine (PS). Long Term Storage: +4°C. Applications: Flow Cytometry .
Catalog No. 50-200-8059