Learn More
Enzo Life Sciences Annexin V (human), (recombinant) (R-PE conjugate) (100tests)
SDP
Supplier:  Enzo Life Sciences ALX209251T100
Encompass_Preferred
R-PE-labelled recombinant human annexin V is suitable for dual staining experiments to detect phosphatidylserine whilst cell-surface protein can be detected with a suitable MAb (FITC-labelled or labelled secondary antibody). The fluorochrome R-PE (phycoerythrin) shows red fluorescence (Ex(max): 495-564nm; Em(max): 576nm). Exhibits anti-phospholipase activity. R-PE-labelled recombinant human annexin V is suitable for dual staining experiments to detect phosphatidylserine whilst cell-surface protein can be detected with a suitable MAb (FITC-labelled). The fluorochrome R-PE (phycoerythrin) shows red fluorescence (Ex(max): 495-564nm; Em(max): 576nm). Exhibits anti-phospholipase activity. Purity: ≥89% (SDS-PAGE, HPLC). MW: ~35.8kDa. Formulation: Liquid. In 50mM TRIS, pH 7.4, containing 100mM sodium chloride, 1% BSA and 0.02% sodium azide. Source: Produced in E. coli. Specificity: Binds to phosphatidylserine (PS). Handling: Protect from light. Do not freeze. Applications: Flow Cytometry .
Catalog No. 50-200-8061