Learn More
Medchemexpress LLC Annonacin | 111035-65-5 | 1 MG
SDP
Catalog No. 5000418342
Encompass_Preferred
Annonacin is an acetylgenin that inhibits the mitochondrial complex pathway and ATPase pumps. It is known to increase tau phosphorylation and exhibits significant killing effects on various cancer cells, including ovarian, cervical, breast, bladder, and skin cancer lines. Annonacin induces apoptosis through Bax and Caspase-3-related pathways.
  • Promotes selective cancer cell death via NKA-dependent and SERCA-dependent pathways.
  • Arrests cancer cells at the G1 phase.
  • Induces apoptosis through Bax and Caspase-3-related pathways.
  • Increases tau phosphorylation.
  • Exhibits anti-cancer effects in mouse models of skin tumors.
  • Has a purity of 98.09%.
Catalog No. 50-004-18342 Supplier Medchemexpress LLC Supplier No. HYN28771MG
Annonacin is an acetylgenin that inhibits the mitochondrial complex pathway and ATPase pumps. It is known to increase tau phosphorylation and exhibits significant killing effects on various cancer cells, including ovarian, cervical, breast, bladder, and skin cancer lines. Annonacin induces apoptosis through Bax and Caspase-3-related pathways.
  • Promotes selective cancer cell death via NKA-dependent and SERCA-dependent pathways.
  • Arrests cancer cells at the G1 phase.
  • Induces apoptosis through Bax and Caspase-3-related pathways.
  • Increases tau phosphorylation.
  • Exhibits anti-cancer effects in mouse models of skin tumors.
  • Has a purity of 98.09%.