Learn More
Electron Microscopy Sciences Annular Sputter Target Gold, 87 mm O.D., 57 mm I.D.
SDP
Catalog No. 5032883
Encompass
Targets have a standard thickness of 0.1 mm (100 micron) or use on most sputtering units and are available mounted or unmounted. Made from high purity metals. When ordering, please state make and model of the sputtering unit to ensure proper fit.
O.D.: 87 mm
I.D.: 57 mm
Catalog No. 50-328-83 Supplier Electron Microscopy Sciences Supplier No. 91031AU
Targets have a standard thickness of 0.1 mm (100 micron) or use on most sputtering units and are available mounted or unmounted. Made from high purity metals. When ordering, please state make and model of the sputtering unit to ensure proper fit.
O.D.: 87 mm
I.D.: 57 mm