Learn More
Aviva Systems Biology Corp ANO6 Antibody -C-terminal region (ARP44761_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44761_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM16F. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C-terminal region of human ANO6. Alternate names: MGC104751, SCTS, BDPLT7, TMEM16FApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54039