Learn More
Crescent Chemical Co Inc ANODE FLUID 3 FOR IEF
SDP
Catalog No. 50248730
Encompass_Preferred
Anode fluid 3 for IEF Contains 0.17 g L-aspartic acid and 0.18 g L-glutamic acid in 50 ml water. Recommended for general use with SERVALYT™ PRECOTES™ Size - 50ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42984.03
Catalog No. 50-248-730 Supplier Crescent Chemical Co Inc Supplier No. SE42984.03
Anode fluid 3 for IEF Contains 0.17 g L-aspartic acid and 0.18 g L-glutamic acid in 50 ml water. Recommended for general use with SERVALYT™ PRECOTES™ Size - 50ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42984.03