Learn More
Thermo Scientific Anode Caps | Stainless Steel | 50 Pc
Supplier:  Thermo Scientific 1187441
Encompass
Stainless steel Anode Caps are disposable items to avoid any carry-over from sample to sample. Graphite parts are designed to be cleaned wet chemically using mineral acids. With the handy removal tool, replacing the Anode Caps is a quick and simple process.
Catalog No. NC2127542