Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Anomalin | 4970-26-7 | 426.46 | 5 MG
SDP

Catalog No. 5000416806
Encompass_Preferred

(-)-Anomalin is a coumarin derivative isolated from the root of Angelica anomala. It is also known as (-)-Praeruptorin B.

  • Purity: 99.81%
  • Appearance: solid
  • Color: white to yellow
  • Structure classification: phenylpropanoids, coumarins
  • Initial source: plants umbelliferae, plants labiatae salvia hypargeia fisch. & c.a.mey., plants compositae artemisia vulgaris l.

Catalog No. 50-004-16806 Supplier Medchemexpress LLC Supplier No. HYN09475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Anomalin is a coumarin derivative isolated from the root of Angelica anomala. It is also known as (-)-Praeruptorin B.

  • Purity: 99.81%
  • Appearance: solid
  • Color: white to yellow
  • Structure classification: phenylpropanoids, coumarins
  • Initial source: plants umbelliferae, plants labiatae salvia hypargeia fisch. & c.a.mey., plants compositae artemisia vulgaris l.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.