Learn More
AdipoGen Anomalin A (1 mg)
SDP
Supplier:  AdipoGen AGCN20006M001
Encompass_Preferred
Chemical. CAS: 548740-86-9. Formula: C14H10O6. MW: 274.1. Isolated from marine fungus Arthrinium sp. Lck (p56lck; lymphocyte specific tyrosine kinase) inhibitor. Antioxidant. DPPH radical scavenger.
Catalog No. 50-205-3352