Learn More
Cayman Chemical Anominine 100ug
SDP
Catalog No. 502947623
Encompass_Preferred
An indole diterpene fungal metabolite; dietary administration induces mortality in 40% of H. zea at 100 ppm
Catalog No. 50-294-7623 Supplier Cayman Chemical Supplier No. 32923100 UG
An indole diterpene fungal metabolite; dietary administration induces mortality in 40% of H. zea at 100 ppm