Learn More
TARGETMOL CHEMICALS INC --ANONAINE 5MG
SDP
Catalog No. 502578895
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (-)-Anonaine can be extracted from several species of Magnoliaceae and Annelidae and has antimalarial antibacterial antifungal antioxidant anticancer antidepressant and vasodilatory activities. (-)-Anonaine induces apoptosis in human cervical cancer (HeLa) cells induces DNA damage and inhibits the growth and migration of human lung cancer h1299 cells through Bax and caspase-dependent pathways. purity: 99%
Catalog No. 50-257-8895 Supplier TARGETMOL CHEMICALS INC Supplier No. TN13935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (-)-Anonaine can be extracted from several species of Magnoliaceae and Annelidae and has antimalarial antibacterial antifungal antioxidant anticancer antidepressant and vasodilatory activities. (-)-Anonaine induces apoptosis in human cervical cancer (HeLa) cells induces DNA damage and inhibits the growth and migration of human lung cancer h1299 cells through Bax and caspase-dependent pathways. purity: 99%