Learn More
Eurofins Discoverx ANP(1-28), 0.5 mg. (92-1337)
Supplier:  Eurofins Discoverx 921337
Encompass
Atrial natriuretic peptide (ANP), a stimulator of particulate guanylate cyclase, has been found to protect against reoxygenation-induced hypercontracture in isolated cardiomyocytes by increasing cyclic guanosine monophosphate synthesis; ANP also contributes to stimulation of lipolysis during repeated bouts of exercise and in the control of lipolysis during exercise. Provided as 0.5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9846