Learn More
Echelon Biosciences Research Labs ANP (126-150), rat / Auriculin B, ANP (4-28), rat - 0.5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 155620.5MG
Encompass_Preferred
ANP (126-150), rat / Auriculin B, ANP (4-28), rat - 0.5mg , Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr-OH This item is non returnable
Catalog No. 50-190-8254