Learn More
Aviva Systems Biology Corp ANP32A Antibody - N-terminal region (ARP40204_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40204_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANP32A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ANP32A. Alternate names: LANP, MAPM, PP32, HPPCn, PHAP1, PHAPI, I1PP2A, C15orf1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57338