Learn More
Sino Biological ANP32A / PHAP1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11942T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ANP32A / PHAP1 (rh ANP32A / PHAP1; Catalog#11942-H20E; NP_006296.1; Glu2-Lys238). ANP32A / PHAP1 specific IgG was purified by Human ANP32A / PHAP1 affinity chromatography.
Catalog No. 50-224-7576