Learn More
Aviva Systems Biology Corp ANPEP Antibody - N-terminal region (ARP61079_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61079_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANPEP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ANPEP. Alternate names: APN, CD13, GP150, LAP1, P150, PEPNApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-287-243