Learn More
Abcam Anpirtoline hydrochloride, 5-HT1B receptor agonist, 10MG
Catalog No. 502704953
Encompass_Preferred
MW 265.2 Da, Purity >99%. Potent, selective 5-HT1B receptor agonist (Ki values are 28, 150 and 1490 nM for 5-HT1B, 5-HT1A and 5-HT2 receptors, respectively). Shows antidyskinetic effects *in vivo*. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4953 Supplier Abcam Supplier No. AB14612210MG
MW 265.2 Da, Purity >99%. Potent, selective 5-HT1B receptor agonist (Ki values are 28, 150 and 1490 nM for 5-HT1B, 5-HT1A and 5-HT2 receptors, respectively). Shows antidyskinetic effects *in vivo*. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement