Learn More
Medchemexpress LLC Anpirtoline hydrochloride | 99201-87-3 | 99.9% | 265.21 | C10H14Cl2N2S | 100 MG
SDP
Catalog No. 5000213320
Encompass_Preferred
Anpirtoline hydrochloride is a research-grade serotonergic compound that functions primarily as a 5-HT1B receptor agonist, with activity at 5-HT1A and 5-HT2 subtypes. It is supplied for laboratory research use only and is not for human or veterinary use. CAS: 99201-87-3; molecular weight: 265.21 g/mol.
  • Potent 5-HT1B receptor agonist with activity at related 5-HT subtypes.
  • High reported purity, suitable for pharmacological experiments.
  • Available in multiple pack sizes to support screening or preparative work.
  • Provided with documentation useful for research-grade handling and QC.
Catalog No. 50-002-13320 Supplier Medchemexpress LLC Supplier No. HY101100100MG
Anpirtoline hydrochloride is a research-grade serotonergic compound that functions primarily as a 5-HT1B receptor agonist, with activity at 5-HT1A and 5-HT2 subtypes. It is supplied for laboratory research use only and is not for human or veterinary use. CAS: 99201-87-3; molecular weight: 265.21 g/mol.
  • Potent 5-HT1B receptor agonist with activity at related 5-HT subtypes.
  • High reported purity, suitable for pharmacological experiments.
  • Available in multiple pack sizes to support screening or preparative work.
  • Provided with documentation useful for research-grade handling and QC.