Learn More
Medchemexpress LLC Anrikefon acetate | 2584931-05-3 | 99.7% | 763.97 | 10 MG
SDP
Catalog No. 5000396201
Encompass_Preferred
Anrikefon acetate is a kappa opioid receptor agonist that exhibits an analgesic effect. This product is strictly for research purposes and is not intended for human or patient use.
  • Kappa opioid receptor agonist
  • Analgesic effect
  • For research use only
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO
Catalog No. 50-003-96201 Supplier Medchemexpress LLC Supplier No. HYP3445A10MG
Add to Cart
Add to Cart
Anrikefon acetate is a kappa opioid receptor agonist that exhibits an analgesic effect. This product is strictly for research purposes and is not intended for human or patient use.
  • Kappa opioid receptor agonist
  • Analgesic effect
  • For research use only
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO