Learn More
TARGETMOL CHEMICALS INC Ansamitocin p-3 10MG
SDP
Catalog No. 502877389
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ansamitocin p-3 (Maytansinol isobutyrate) is an inhibitor of microtubule with IC50 of 3.4 uM for tubulin polymerization. Purity 99.73%
Catalog No. 50-287-7389 Supplier TARGETMOL CHEMICALS INC Supplier No. T2226210MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ansamitocin p-3 (Maytansinol isobutyrate) is an inhibitor of microtubule with IC50 of 3.4 uM for tubulin polymerization. Purity 99.73%