Learn More
Medchemexpress LLC HY-15739 100mg , Ansamitocin P-3 Antibiotic C 15003P3;Maytansinol butyrate;C15003P3 CAS:66584-72-3 Purity:98%
SDP
Catalog No. 501493113
Encompass_Preferred
Medchemexpress, HY-15739 100mg Ansamitocin P-3 Antibiotic C 15003P3;Maytansinol butyrate;C15003P3 CAS:66584-72-3 Formula:C32H43ClN2O9 Microtubule Purity:98% Ansamitocin P-3 is a microtubule inhibitor. Ansamitocin P-3 is a macrocyclic antitumor antibiotic. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3113 Supplier Medchemexpress LLC Supplier No. HY15739100MG
Medchemexpress, HY-15739 100mg Ansamitocin P-3 Antibiotic C 15003P3;Maytansinol butyrate;C15003P3 CAS:66584-72-3 Formula:C32H43ClN2O9 Microtubule Purity:98% Ansamitocin P-3 is a microtubule inhibitor. Ansamitocin P-3 is a macrocyclic antitumor antibiotic. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.