Learn More
Selleck Chemical LLC Ansamitocin p-3 (Maytansinol isobutyrate. NSC292222) S2447-5mg
SDP
Supplier:  Selleck Chemical LLC S24475MG
Encompass_Preferred
Ansamitocin p-3 (Maytansinol isobutyrate NSC292222 Antibiotic C 15003P3) is a potent inhibitor of tubulin polymerization with IC50 of 3 4 M
Catalog No. 50-311-1920