Learn More
Medchemexpress LLC Ansamitocin P-3 (Antibiotic C 15003P3) | 66584-72-3 | MFCD00274586 | 98.0% | 635.14 | C32H43ClN2O9 | 10 MG
SDP
Catalog No. 5000288412
Encompass_Preferred
Ansamitocin P-3 is a maytansinoid-class macrocyclic antitumor antibiotic and potent microtubule/tubulin inhibitor supplied for research use as a solid. It is commonly used as a cytotoxic payload for antibody-drug conjugate research and for in vitro studies of microtubule disruption and cancer cell cytotoxicity.
  • Potent microtubule/tubulin inhibitor suitable for cytotoxicity and ADC research.
  • Maytansinoid-class payload with demonstrated activity against multiple cancer cell lines.
  • Provided in milligram-scale quantities (1 mg, 5 mg, 10 mg) for laboratory use.
  • Soluble in DMSO at ≥ 100 mg/mL; compatible with several in vivo vehicle preparations.
  • Stable as a powder at -20°C for up to three years; follow recommended in-solution storage conditions.
Catalog No. 50-002-88412 Supplier Medchemexpress LLC Supplier No. HY1573910MG
Ansamitocin P-3 is a maytansinoid-class macrocyclic antitumor antibiotic and potent microtubule/tubulin inhibitor supplied for research use as a solid. It is commonly used as a cytotoxic payload for antibody-drug conjugate research and for in vitro studies of microtubule disruption and cancer cell cytotoxicity.
  • Potent microtubule/tubulin inhibitor suitable for cytotoxicity and ADC research.
  • Maytansinoid-class payload with demonstrated activity against multiple cancer cell lines.
  • Provided in milligram-scale quantities (1 mg, 5 mg, 10 mg) for laboratory use.
  • Soluble in DMSO at ≥ 100 mg/mL; compatible with several in vivo vehicle preparations.
  • Stable as a powder at -20°C for up to three years; follow recommended in-solution storage conditions.